accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 769-30-2

OOOH

CAS RN 769-30-2

IUPAC Name: (2H-1,3-benzodioxol-4-yl)methanol
Synonyms: 2H-1,3-benzodioxol-4-ylmethanol
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: XVCMMPXFVAHHQN-UHFFFAOYSA-N
SMILES: OCC1=C2OCOC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results