accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 769-42-6

IUPAC Name: 1,3-dimethyl-1,3-diazinane-2,4,6-trione
Synonyms: 1,3-dimethyl-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol): 156.14
Molecular Formula: C6H8N2O3
InChi Key: VVSASNKOFCZVES-UHFFFAOYSA-N
SMILES: CN1C(=O)CC(=O)N(C)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)