accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 769-78-8

IUPAC Name: ethenyl benzoate
Synonyms: benzoic acid, vinyl ester
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: KOZCZZVUFDCZGG-UHFFFAOYSA-N
SMILES: C=COC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)