accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7691-02-3

IUPAC Name: bis(ethenyldimethylsilyl)amine
Synonyms: bis(ethenyldimethylsilyl)amine
Molecular Weight (g/mol): 185.42
Molecular Formula: C8H19NSi2
InChi Key: WYUIWUCVZCRTRH-UHFFFAOYSA-N
SMILES: C[Si](C)(N[Si](C)(C)C=C)C=C
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)