accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7693-41-6

IUPAC Name: 4-methoxyphenyl carbonochloridate
Synonyms: 4-methoxyphenyl carbonochloridate
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: CCFSGQKTSBIIHG-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(Cl)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)