accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7693-44-9

OOClBr

CAS RN 7693-44-9

IUPAC Name: 4-bromophenyl carbonochloridate
Synonyms: 4-bromophenyl carbonochloridate
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: IKMNJYGTSSQNSE-UHFFFAOYSA-N
SMILES: ClC(=O)OC1=CC=C(Br)C=C1
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results