accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7696-12-0

IUPAC Name: (1,3-dioxo-2,3,4,5,6,7-hexahydro-1H-isoindol-2-yl)methyl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Synonyms: tetramethrin
Molecular Weight (g/mol): 331.41
Molecular Formula: C19H25NO4
InChi Key: CXBMCYHAMVGWJQ-CABCVRRESA-N
SMILES: CC(C)=C[C@@H]1[C@@H](C(=O)OCN2C(=O)C3=C(CCCC3)C2=O)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results