CAS RN 7697-29-2
IUPAC Name:
4-chloro-3-methylbenzoic acid
Synonyms:
4-chloro-3-methylbenzoic acid
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
MRUKIIWRMSYKML-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=C1Cl)C(O)=O