accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76981-71-0

IUPAC Name: ethyl (6S)-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Synonyms: ethyl (6S)-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular Weight (g/mol): 239.33
Molecular Formula: C12H17NO2S
InChi Key: WSOISFMHBFFWRQ-ZETCQYMHSA-N
SMILES: CCOC(=O)C1=C(N)SC2=C1CC[C@H](C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results