accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COONH(R)OHO

CAS RN 76985-10-9

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(naphthalen-2-yl)propanoic acid| BOC-3-(2-NAPHTHYL)-D-ALANINE
Molecular Weight (g/mol): 315.37
Molecular Formula: C18H21NO4
InChi Key: URKWHOVNPHQQTM-OAHLLOKOSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)