accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77-26-9

IUPAC Name: 5-(2-methylpropyl)-5-(prop-2-en-1-yl)-1,3-diazinane-2,4,6-trione
Synonyms: butalbital butalbital sodium allylbarbituric acid 布他比妥 allylbarbitone allylbarbital butalbitalum butalbarbital tetrallobarbital butalbital M (OH)
Molecular Weight (g/mol): 224.26
Molecular Formula: C11H16N2O3
InChi Key: UZVHFVZFNXBMQJ-UHFFFAOYSA-N
SMILES: CC(C)CC1(CC=C)C(=O)NC(=O)NC1=O
Concentration 
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)