CAS RN 77-36-1
IUPAC Name:
2-chloro-5-(1-hydroxy-3-oxo-2,3-dihydro-1H-isoindol-1-yl)benzene-1-sulfonamide
Synonyms:
chlorthalidone oksodolin (oxodolin) igroton chlortalidone (inn) (+-)-chlorthalidone oxodoline chlorphthalidolone zambesil phthalamodine hydro-long
Molecular Weight (g/mol):
338.76
Molecular Formula:
C14H11ClN2O4S
InChi Key:
JIVPVXMEBJLZRO-UHFFFAOYNA-N
SMILES:
NS(=O)(=O)C1=CC(=CC=C1Cl)C1(O)NC(=O)C2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
Chlorthalidone, 98%
Chlorthalidone, 98%
CAS: 77-36-1 Molecular Formula: C14H11ClN2O4S Molecular Weight (g/mol): 338.77 InChI Key: JIVPVXMEBJLZRO-UHFFFAOYSA-N Synonym: chlorthalidone,chlortalidone,phthalamudine,chlorphthalidolone,hygroton,chlorthalidon,phthalamodine,chlorothalidone,chlorphthalidone,natriuran PubChem CID: 2732 ChEBI: CHEBI:3654 IUPAC Name: 2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide SMILES: C1=CC=C2C(=C1)C(=O)NC2(C3=CC(=C(C=C3)Cl)S(=O)(=O)N)O