accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)(S)H3CCH3(R)(S)(R)CH3OH

CAS RN 77-53-2

IUPAC Name: (1S,2R,5S,7R,8R)-2,6,6,8-tetramethyltricyclo[5.3.1.0¹,⁵]undecan-8-ol
Synonyms: eudesmol (8R)-cedran-8-ol cedrol (natural) α-cedrol 8betah-cedran-8-ol 8-betah-cedran-8-ol (8CI) cedrol 8betah-cedran-8-ol (8CI) cedarwood oil alcohols (+)-cedrol
Molecular Weight (g/mol): 222.37
Molecular Formula: C15H26O
InChi Key: SVURIXNDRWRAFU-OGMFBOKVSA-N
SMILES: C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)