accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77-71-4

IUPAC Name: 5,5-dimethylimidazolidine-2,4-dione
Synonyms: DM hydantoin
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: YIROYDNZEPTFOL-UHFFFAOYSA-N
SMILES: CC1(C)NC(=O)NC1=O
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)