accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77-79-2

IUPAC Name: 2,5-dihydro-1λ⁶-thiophene-1,1-dione
Synonyms: sulfolene
Molecular Weight (g/mol): 118.15
Molecular Formula: C4H6O2S
InChi Key: MBDNRNMVTZADMQ-UHFFFAOYSA-N
SMILES: O=S1(=O)CC=CC1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (2)
  • (9)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (7)
  • (8)
  • (3)
  • (2)
  • (9)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)