CAS RN 77-92-9
Citric acid, anhydrous, ACS, 99.5+%
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric acid, 99+%
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric acid, ≥99.5%, ACS reagent, anhydrous
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric acid, for molecular biology, anhydrous, Rnase and Protease free
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid, Anhydrous (Crystalline/USP), Fisher Chemical™
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid Anhydrous (Crystalline), Fisher BioReagents
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric acid, 99%, pure, anhydrous
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric acid, Anhydrous, 99.5 to 100.5% (Dry Basis)
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.123 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: C(C(=O)O)C(CC(=O)O)(C(=O)O)O
Citric acid, 99.5%, for analysis, anhydrous
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid, Anhydrous, 99.5 to 100.5%, MilliporeSigma™
CAS: 77-92-9 Molecular Formula: C6H8O7 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid Electrophoresis Grade, 1kg, MP Biomedicals
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid, Powder, Multi-Compendial, J.T. Baker™
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid, Anhydrous, Granular, FCC, 99.5-100.5%, Spectrum™ Chemical
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O
Citric Acid, Powder, U.S.P., J.T. Baker™
CAS: 77-92-9 Molecular Formula: C6H8O7 Molecular Weight (g/mol): 192.12 MDL Number: MFCD00011669 InChI Key: KRKNYBCHXYNGOX-UHFFFAOYSA-N Synonym: citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy PubChem CID: 311 ChEBI: CHEBI:30769 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O