CAS RN 77026-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(±)-Jasmonic acid, ≥90% (GC), MilliporeSigma™ Supelco™
(+/-)-Jasmonic acid is a plant lipid derivative and a naturally available growth regulator found in higher plants.