accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrBr

CAS RN 7703-74-4

IUPAC Name: 2,6-bis(bromomethyl)pyridine
Synonyms: 2,6-bis(bromomethyl)pyridine
Molecular Weight (g/mol): 264.95
Molecular Formula: C7H7Br2N
InChi Key: QUTSYCOAZVHGGT-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC(CBr)=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)