accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClClSN

CAS RN 7709-58-2

IUPAC Name: hydrogen 4-(chloromethyl)-1,3-thiazole chloride
Synonyms: hydrogen 4-(chloromethyl)-1,3-thiazole chloride
Molecular Weight (g/mol): 170.05
Molecular Formula: C4H5Cl2NS
InChi Key: NVTBASMQHFMANH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCC1=CSC=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)