accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 771-99-3

IUPAC Name: 4-phenylpiperidine
Synonyms: 4-phenylpiperidine
Molecular Weight (g/mol): 161.25
Molecular Formula: C11H15N
InChi Key: UTBULQCHEUWJNV-UHFFFAOYSA-N
SMILES: C1CC(CCN1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Quantity 
  • (4)
  • (3)
Physical Form 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)