accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77113-48-5

IUPAC Name: (5-chloropent-1-yn-1-yl)trimethylsilane
Synonyms: (5-chloropent-1-yn-1-yl)trimethylsilane
Molecular Weight (g/mol): 174.74
Molecular Formula: C8H15ClSi
InChi Key: UPVHIBLXQNNITH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CCCCCl
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results