accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3O

CAS RN 77123-58-1

IUPAC Name: 2-[2-(trimethylsilyl)ethynyl]benzaldehyde
Synonyms: 2-[2-(trimethylsilyl)ethynyl]benzaldehyde
Molecular Weight (g/mol): 202.33
Molecular Formula: C12H14OSi
InChi Key: IROGCMISRBINQX-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results