accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOHOOH2O

CAS RN 77128-70-2

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)acetic acid hydrate
Synonyms: {[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}acetic acid hydrate
Molecular Weight (g/mol): 329.35
Molecular Formula: C18H19NO5
InChi Key: CUJSWOOWOONPRH-UHFFFAOYSA-N
SMILES: O.CN(CC(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)