accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHNHOO

CAS RN 77128-72-4

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methoxyphenyl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 417.46
Molecular Formula: C25H23NO5
InChi Key: JYQODLWFOPCSCS-UHFFFAOYNA-N
SMILES: COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results