accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7713-71-5

IUPAC Name: methyl(1-phenylpropyl)amine
Synonyms: methyl(1-phenylpropyl)amine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: YELGXBUYFOSFJM-UHFFFAOYNA-N
SMILES: CCC(NC)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results