accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrBr

CAS RN 77143-76-1

IUPAC Name: ethyl 2-bromo-2-(4-bromophenyl)acetate
Synonyms: ethyl 2-bromo-2-(4-bromophenyl)acetate
Molecular Weight (g/mol): 322
Molecular Formula: C10H10Br2O2
InChi Key: PPRAWWBOUOBAON-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)C1=CC=C(Br)C=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)