accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 771572-29-3

IUPAC Name: 1-(5-methyl-2-phenylfuran-3-yl)methanamine
Synonyms: 1-(5-methyl-2-phenylfuran-3-yl)methanamine
Molecular Weight (g/mol): 187.24
Molecular Formula: C12H13NO
InChi Key: PQZVRVFUCJMCRZ-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results