CAS RN 77167-06-7
CAS RN 77167-06-7
IUPAC Name:
N-(2,6-dichlorophenyl)benzenesulfonamide
Synonyms:
N-(2,6-dichlorophenyl)benzenesulfonamide
Molecular Weight (g/mol):
302.17
Molecular Formula:
C12H9Cl2NO2S
InChi Key:
CQELSZQJFRLUFQ-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(Cl)=C1NS(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals N-(2,6-Dichlorophenyl)benzenesulfonamide, 97%, Thermo Scientific™
CAS: 77167-06-7 Molecular Formula: C12H9Cl2NO2S Molecular Weight (g/mol): 302.169 MDL Number: MFCD02007023 InChI Key: CQELSZQJFRLUFQ-UHFFFAOYSA-N Synonym: n-2,6-dichlorophenyl benzenesulfonamide PubChem CID: 12723885 IUPAC Name: N-(2,6-dichlorophenyl)benzenesulfonamide SMILES: C1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC=C2Cl)Cl