accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7718-98-1

IUPAC Name: vanadium(3+) trichloride
Synonyms: vanadium(III) chloride
Molecular Weight (g/mol): 157.29
Molecular Formula: Cl3V
InChi Key: HQYCOEXWFMFWLR-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[V+3]
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (5)
  • (5)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)