accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 772-00-9

IUPAC Name: 4-phenyl-1,3-dioxane
Synonyms: 1,3-dioxane, 4-phenyl-
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: RCJRILMVFLGCJY-UHFFFAOYNA-N
SMILES: C1CC(OCO1)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)