accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3CH

CAS RN 772-38-3

IUPAC Name: 1-tert-butyl-4-ethynylbenzene
Synonyms: 1-tert-butyl-4-ethynylbenzene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: ZSYQVVKVKBVHIL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C#C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)