accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3N

CAS RN 77200-12-5

IUPAC Name: N-ethyl-N-methylpyridin-2-amine
Synonyms: N-ethyl-N-methylpyridin-2-amine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: MKTSSBCZDAXGGO-UHFFFAOYSA-N
SMILES: CCN(C)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results