accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77253-67-9

IUPAC Name: trifluoro(²H₂)ethan-1-(²H)ol
Synonyms: trifluoro(²H₂)ethan(²H)ol
Molecular Weight (g/mol): 103.06
Molecular Formula: C2H3F3O
InChi Key: RHQDFWAXVIIEBN-IDPMSXFZSA-N
SMILES: [2H]OC([2H])([2H])C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results