accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77257-42-2

IUPAC Name: triethyl(2-{4-[(1E)-2-phenylethenyl]phenoxy}ethyl)azanium iodide
Synonyms: triethyl(2-{4-[(1E)-2-phenylethenyl]phenoxy}ethyl)azanium iodide
Molecular Weight (g/mol): 451.39
Molecular Formula: C22H30INO
InChi Key: RDTKUZXIHMTSJO-UEIGIMKUSA-M
SMILES: [I-].CC[N+](CC)(CC)CCOC1=CC=C(\C=C\C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results