accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOOCH3

CAS RN 7726-20-7

IUPAC Name: 1-(octane-1-sulfonyl)octane
Synonyms: 1-(octane-1-sulfonyl)octane
Molecular Weight (g/mol): 290.51
Molecular Formula: C16H34O2S
InChi Key: TZPCGEVFZZYCIY-UHFFFAOYSA-N
SMILES: CCCCCCCCS(=O)(=O)CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)