CAS RN 7726-95-6
Bromine Water (Saturated at 20°C/Reagent), Fisher Chemical
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 MDL Number: MFCD00010896 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 IUPAC Name: bromide SMILES: [Br-]
Bromine liquid, 99.8%
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 MDL Number: MFCD00010896 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 SMILES: [Br-]
Bromine, 99.5+%, ACS reagent
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 IUPAC Name: molecular bromine SMILES: [Br-]
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
CAS: 7726-95-6 | Molecular Formula: Br | Molecular Weight: 79.91 g/mol
Bromine, 99.5+%, ACS reagent
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 IUPAC Name: molecular bromine SMILES: [Br-]
Bromine, 99+%, extra pure
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 MDL Number: MFCD00010896 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 SMILES: [Br-]
Bromine liquid, ACS, 99.5%
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 MDL Number: MFCD00010896 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 SMILES: [Br-]
Bromine liquid, 99.99+% (metals basis)
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 MDL Number: MFCD00010896 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 SMILES: [Br-]
Bromine Water, Saturated Solution, Spectrum™ Chemical
Bromine, 99.6%, for analysis
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 MDL Number: MFCD00010896 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 SMILES: [Br-]
Bromine, 1M solution in acetic acid, AcroSeal™
CAS: 7726-95-6 | Br | 79.91 g/mol
Bromine Water, Saturated, approximately 3% (w/v), Ricca Chemical
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 IUPAC Name: bromide SMILES: [Br-]
Bromine concentrate, For 1L standard solution, 0.05 M Br2, Solstice
CAS: 7726-95-6 Molecular Formula: Br Molecular Weight (g/mol): 79.91 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-M Synonym: bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish PubChem CID: 24408 ChEBI: CHEBI:29224 IUPAC Name: molecular bromine SMILES: [Br-]
Sigma Aldrich Allo-Inositol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2