accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClVClClO

CAS RN 7727-18-6

IUPAC Name: oxovanadiumpentakis(ylium) trichloride
Synonyms: oxovanadiumpentakis(ylium) trichloride
Molecular Weight (g/mol): 173.29
Molecular Formula: Cl3OV
InChi Key: WMZFXRASJAIALG-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].O=[V+5]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results