accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COIO

CAS RN 77287-58-2

IUPAC Name: 2-iodo-5-methoxybenzaldehyde
Synonyms: 2-iodo-5-methoxybenzaldehyde
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: YHRFMOFYYPMQBE-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results