accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77297-00-8

IUPAC Name: bis(N,N-diethylguanidine); sulfuric acid
Synonyms: bis(N,N-diethylguanidine); sulfuric acid
Molecular Weight (g/mol): 328.43
Molecular Formula: C10H28N6O4S
InChi Key: VZAFCEFGYOMBML-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CCN(CC)C(N)=N.CCN(CC)C(N)=N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results