accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COS

CAS RN 77298-24-9

IUPAC Name: [(2-methoxyethyl)sulfanyl]benzene
Synonyms: [(2-methoxyethyl)sulfanyl]benzene
Molecular Weight (g/mol): 168.25
Molecular Formula: C9H12OS
InChi Key: ONVZBVAUSAELNQ-UHFFFAOYSA-N
SMILES: COCCSC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results