accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH

CAS RN 7731-28-4

IUPAC Name: 4-methylcyclohexan-1-ol
Synonyms: 4-methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: MQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILES: CC1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results