accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77359-11-6

IUPAC Name: 3-[(4-nitrophenyl)methoxy]-3-oxopropanoic acid
Synonyms: 3-[(4-nitrophenyl)methoxy]-3-oxopropanoic acid
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: RIGFMUNSTCPGNP-UHFFFAOYSA-N
SMILES: OC(=O)CC(=O)OCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results