accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7737-62-4

IUPAC Name: ethyl 3-oxoheptanoate
Synonyms: ethyl 3-oxoheptanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: UKRVECBFDMVBPU-UHFFFAOYSA-N
SMILES: CCCCC(=O)CC(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results