accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHCH3H3CCH3

CAS RN 77387-57-6

IUPAC Name: (3,5-di-tert-butylphenyl)methanol
Synonyms: (3,5-di-tert-butylphenyl)methanol
Molecular Weight (g/mol): 220.36
Molecular Formula: C15H24O
InChi Key: VHYHRNYPVNFGNR-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(CO)=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results