accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOF

CAS RN 773872-57-4

IUPAC Name: {2'-fluoro-[1,1'-biphenyl]-4-yl}methanol
Synonyms: {2'-fluoro-[1,1'-biphenyl]-4-yl}methanol
Molecular Weight (g/mol): 202.23
Molecular Formula: C13H11FO
InChi Key: WABWIEVUZWQZLC-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results