accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 774-48-1

IUPAC Name: (diethoxymethyl)benzene
Synonyms: (diethoxymethyl)benzene
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: MAQMEXSLUSZDQM-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results