accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 774-53-8

IUPAC Name: 5-bromo-2,3'-bipyridine
Synonyms: 5-bromo-2,3'-bipyridine
Molecular Weight (g/mol): 235.08
Molecular Formula: C10H7BrN2
InChi Key: ILJRIAPZWHYPMK-UHFFFAOYSA-N
SMILES: BrC1=CN=C(C=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results