accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 774-55-0

IUPAC Name: 1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Synonyms: 1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: VEPUKHYQNXSSKV-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2CCCCC2=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)