accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 774-65-2

IUPAC Name: tert-butyl benzoate
Synonyms: tert-butyl benzoate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: LYDRKKWPKKEMNZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results