accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSFFFFF

CAS RN 774-94-7

IUPAC Name: 4-(pentafluoro-λ⁶-sulfanyl)phenol
Synonyms: 4-(pentafluorosulfanyl)phenol
Molecular Weight (g/mol): 220.16
Molecular Formula: C6H5F5OS
InChi Key: XHJLGVIUMCBMHL-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results